CPC® High-Flow Quick-Disconnect Fittings, Elbow Hose Barb Inserts
Supplier: VWR International
MFLX31303-51 MFLX31303-55 MFLX31303-56 MFLX31307-56 HFC23812 (10PK) HFCD23612 (10PK) HFCD23812 (10PK) HFCD23835 (10PK) HFC23835
MFLX31303-50EA 20.7 CAD
MFLX31303-50 MFLX31303-51 MFLX31303-55 MFLX31303-56 MFLX31307-55 MFLX31307-56 MFLX31308-75 MFLX31303-51BLK MFLX31303-55BLK MFLX31303-56BLK MFLX31307-56BLK MFLX31308-75BLK
CPC® High-Flow Quick-Disconnect Fittings, Elbow Hose Barb Inserts
Fittings and Connectors
No more leaks—valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many 1/2" flow couplings in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. ToMore Product Information