CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Bodies
Supplier: CPC (Colder Products Company)
HFCD171235 (1/PK) HFC171012 HFC171235 HFCD17635 (10PK) HFC17835 HFC17835 HFCD171212 (10PK) HFC17635 (1/PK) HFC17635 (10PK) HFCD17635 (1/PK) HFCD17835 (10PK) HFCD171235 (10PK) HFCD171012 (1/PK) HFCD171012 (10PK) HFC17812 (10PK) HFCD17812 (1/PK) HFCD17835 (1/PK) HFCD17612 (10PK) HFC17612 (10PK) HFC171212 HFC171012 HFCD171212 (1/PK) HFCD17612 (1/PK) HFC17812 (1/PK) HFCD17812 (10PK) HFC17612 (1/PK)
MFLX31303-12BLKEA 271.76 CAD
MFLX31303-12BLK MFLX31303-16BLK MFLX31307-15BLK MFLX31308-34 MFLX31303-10BLK MFLX31307-10BLK MFLX31303-18 MFLX31307-18 MFLX31303-13 MFLX31303-11BLK MFLX31303-15 MFLX31303-13BLK MFLX31303-16 MFLX31308-63BLK MFLX31303-15BLK MFLX31307-13 MFLX31303-10 MFLX31303-11 MFLX31307-15 MFLX31303-12 MFLX31307-10 MFLX31308-34BLK MFLX31307-11 MFLX31308-63 MFLX31307-11BLK MFLX31307-13BLK
CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Bodies
Fittings and Connectors
No more leaks—valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many 1/2" flow couplings in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.