CPC® High-Flow Quick-Disconnect Fittings, Threaded Bodies
Supplier: VWR International
HFCD10612 (1/PK) HFCD10812 (1/PK) HFCD101212 (1/PK) HFC10812 (1/PK) HFCD10635 (1/PK) HFCD10835 (1/PK) HFCD101235 (1/PK) HFCD10612 (10PK) HFCD10812 (10PK) HFCD101212 (10PK) HFC10812 (10PK) HFCD10635 (10PK) HFCD10835 (10PK) HFCD101235 (10PK) HFC10612
MFLX31303-00EA 39.97 CAD
MFLX31303-00 MFLX31303-01 MFLX31303-02 MFLX31303-06 MFLX31303-07 MFLX31307-00 MFLX31307-01 MFLX31307-02 MFLX31308-29 MFLX31308-59 MFLX31303-00BLK MFLX31303-01BLK MFLX31303-02BLK MFLX31303-06BLK MFLX31307-00BLK MFLX31307-01BLK MFLX31307-02BLK MFLX31308-29BLK
CPC® High-Flow Quick-Disconnect Fittings, Threaded Bodies
Fittings and Connectors
No more leaks—valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many 1/2" flow couplings in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.