CPC® Plastic Quick-Disconnect Fittings, Complete Couplings
Supplier: VWR International
APC04 (5PK) APC06 (5PK) 1044200 (10PK) 1044300 (10PK) 1044100 (10PK) APC04 (25PK) APC06 (25PK) 1044200 (250PK) 1044300 (250PK) 1044100 (250PK)
MFLX06360-90EA 47.08 CAD
MFLX06360-90 MFLX06360-91 MFLX31305-00 MFLX31305-05 MFLX31306-05 MFLX06360-90BLK MFLX06360-91BLK MFLX31305-00BLK MFLX31305-05BLK MFLX31306-05BLK
CPC® Plastic Quick-Disconnect Fittings, Complete Couplings
Fittings and Connectors
Fittings connect in a snap.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Acetal fittings are ideal for use with corrosive fluids
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
Complete couplings include one fitting insert and one fitting body.