CPC® Quick-Disconnect Fittings, Compression Inserts
Supplier: VWR International
MFLX06360-30 MFLX06360-41 MFLX06360-74 MFLX06360-75 MFLX06360-77 MFLX06361-32 MFLX06364-30 PMC200412 MFLX06364-75 MFLX31307-53 MFLX31307-54 PMCD2004 (25PK) PMCD20025 (25PK) PLCD20004 (25PK) PLCD20006 (25PK) PLC20006 (25PK) PMCD4004 (25PK) PMCD200412 (25PK) PLCD2000612 (25PK) PLCD40006 PLC40006
MFLX06360-30EA 21.33 CAD
MFLX06360-30 MFLX06360-32 MFLX06360-35 MFLX06360-37 MFLX06360-41 MFLX06360-74 MFLX06360-75 MFLX06360-76 MFLX06360-77 MFLX06361-32 MFLX06364-30 MFLX06364-32 MFLX06364-75 MFLX31307-08 MFLX31307-44 MFLX31307-53 MFLX31307-54 MFLX06360-30BLK MFLX06360-41BLK MFLX06360-74BLK MFLX06360-75BLK MFLX06360-77BLK MFLX06361-32BLK MFLX06364-30BLK MFLX06364-75BLK MFLX31307-54BLK MFLX31307-44BLK
CPC® Quick-Disconnect Fittings, Compression Inserts
Fittings and Connectors
No more struggling—just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).