CPC® Quick-Disconnect Fittings, Push-to-Connect Coupling Bodies
Supplier: VWR International
MFLX31304-00 MFLX31304-02 MFLX31304-10 MFLX31304-50 PLCD14004 (25PK) PLCD14006 (25PK) PLC14004 (25PK) PLCD11004 (25PK) PLC11004
MFLX31304-00EA 39.97 CAD
MFLX31304-00 MFLX31304-02 MFLX31304-10 MFLX31304-50 MFLX31308-55 MFLX31308-56 MFLX31304-00BLK MFLX31304-02BLK MFLX31304-10BLK MFLX31304-50BLK MFLX31308-56BLK
CPC® Quick-Disconnect Fittings, Push-to-Connect Coupling Bodies
Fittings and Connectors
No more struggling—just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-More Product Information