CPC® Miniature Quick-Disconnect Fittings, Panel-Mount Hose Barb Bodies
Supplier: VWR International
MFLX06363-68 MFLX06363-67 MFLX06363-69 MFLX06363-78 MFLX06363-97 SMFPM02 (50PK) SMFPMM3 SMFPM0112 50PK
MFLX06363-66EA 86.43 CAD
MFLX06363-66 MFLX06363-68 MFLX06363-76 MFLX06363-67 MFLX06363-69 MFLX06363-78 MFLX06363-97 MFLX06363-68BLK MFLX06363-97BLK MFLX06363-76BLK
CPC® Miniature Quick-Disconnect Fittings, Panel-Mount Hose Barb Bodies
Fittings and Connectors
No more kinked tubing—allows tubing to rotate freely.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
  • Tubing can rotate freely when connected - preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle 1/16" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.More Product Information