CPC® Miniature Quick-Disconnect Fittings, Hose Barb Bodies
Supplier: VWR International
MFLX06363-54 MFLX06363-55 MFLX06363-56 MFLX06363-57 MFLX06363-64 MFLX06363-61 MFLX06363-59 SMF01 (50PK) SMFD01 (50PK) SMF02 (50PK) SMFD02 (50PK) SMF0212 (50PK)
MFLX06363-54EA 53.09 CAD
MFLX06363-54 MFLX06363-55 MFLX06363-56 MFLX06363-57 MFLX06363-62 MFLX06363-64 MFLX06363-86 MFLX06363-61 MFLX06363-59 MFLX06363-54BLK MFLX06363-55BLK MFLX06363-56BLK MFLX06363-57BLK MFLX06363-64BLK
CPC® Miniature Quick-Disconnect Fittings, Hose Barb Bodies
Fittings and Connectors
No more kinked tubing—allows tubing to rotate freely.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
  • Tubing can rotate freely when connected - preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle 1/16" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.
For a complete coupling, order one fitting body and one insert.