Single Open-End Absolute-Rated Cartridge Systems
Supplier: VWR International
FAP101EGE/1189197 FGF031EGS/1189066 FAP011EGE/1188933 MFE911EGE/1187959 MNY921EGS/1222470 MFE921EGE/1224278 FAP051EGE/1189131 MNY941EGS/1222472 MMP921EGE-E/1244773 MFE941EGE/1227911 FAP031EGE/1189065
MFLX06479-54EA 665.18 CAD
MFLX06479-54 MFLX06479-42 MFLX06479-34 MFLX06479-22 MFLX06479-58 MFLX06479-46 MFLX06479-38 MFLX06479-26 MFLX06479-18 MFLX06479-50 MFLX06479-62
Single Open-End Absolute-Rated Cartridge Systems
Filters Filter Cartridges
A collection to fit a wide range of critical applications.
  • Absolute-rated for use in many regulated processes
  • Meets a wide range of industry and government standards
These SOE cartridges have a 222 O-ring configuration to create a tight seal and prevent influent bypass. They are autoclavable to 250 °F (121 °C) and in-line sterilizable to 257 °F (125 °C) for up to 30-minute cycles; tolerant to an accumulated exposure of 10 hours. The filters may also be sanitized with compatible chemical agents. All cartridges are suitable to 180 °F (82 °C) at 10 psid. Use a SOE cartridge-sump adapter kit (sold separately) to secure cartridges within housings.
The cartridges comply with US CFR Title 21 guidelines for repeated food contact, comply with USP Class VI-121 °C Plastics guidelines and pass the MEM Elution Cytotoxicity Test. Aqueous extracts contain less than 0.25 EU/mL and typically exhibit low levels of non-volatile residues.