Ultrapure Application Double-Open-End Filter Cartridges
Supplier: HARRINGTON INDUSTRIAL PLASTICS LLC
MNY921AAS,9.824"/1228705 FAP011AAE,9.824"/1227998 FAP051AAE, 9.824"/1227278 FAP101AAE, 9.824"/1228121 MFE911AAE, 9.824"/1228707 MFE921AAE, 9.824"/1228923 MMP921AAE-E,9.824"/1244619 MFLX06479-20 MFLX06479-36
MFLX06479-16EA 437.66 CAD
MFLX06479-16 MFLX06479-32 MFLX06479-40 MFLX06479-44 MFLX06479-48 MFLX06479-52 MFLX06479-60 MFLX06479-20 MFLX06479-36
Ultrapure Application Double-Open-End Filter Cartridges
Filters Filter Cartridges
Ideal for ultrapure filtration applications.
  • U.S. CFR Title 21 compliant
These cartridges are autoclavable to 250 °F <121 °C) and in-line sterilizable to 257 °F (125 °C) for up to 30-minute cycles. They are tolerant to an accumulated exposure of 10 hours. The filters may also be sanitized with compatible chemical agents. All cartridges are suitable to 180 °F (82 °C) at 10 psid.
The cartridges comply with U.S. CFR Title 21 guidelines for repeated food contact, comply with USP Class VI, 121 °C Plastics guidelines, and pass the MEM Elution CytotoxicityMore Product Information