Diamond® SureFlow™ Bottle Top Dispensers, Globe Scientific
Supplier: Globe Scientific
GBTD-2.5 GBTD-05 GBTD-10 GBTD-25 GBTD-30 GBTD-50 GBTD-60 GBTD-100 GBTD-R-2.5 GBTD-R-05 GBTD-R-10 GBTD-R-25 GBTD-R-30 GBTD-R-50 GBTD-R-60 GBTD-R-100
76520-454EA 448.59 CAD
76520-454 76520-456 76520-458 76520-460 76520-462 76520-464 76520-466 76520-468 76520-470 76520-472 76520-474 76520-476 76520-478 76520-480 76520-482 76520-484
Diamond® SureFlow™ Bottle Top Dispensers, Globe Scientific
Bottle Top Dispensers
Diamond® SureFlow™ Bottle Top Dispensers have been designed with several features to make the dispensing of valuable laboratory reagents easier than ever.
  • Two-year warranty
  • Fully autoclavable
  • Single tool recalibration
  • Locking feature to prevent spills
  • 360° rotation to dispense in multiple vessels
  • Lower pressure for larger volumes helps reduce splashing
  • Telescopic fill tube for bottles ranging in height from 170 to 300 mm
With a choice of standard or recirculating models, bottle top dispensers include a unique piston design which allows cleaning andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable