VWR® Mini Vortex Mixer

Supplier: VWR International
76549-932
76549-932EA 251.22 CAD
76549-932
VWR® Mini Vortex Mixer
Shakers Vortex Mixers
VWR® mini vortex mixer with standard cup head.

  • Extremely compact design
  • Powerful mixing for tubes up to 50 ml
  • One handed, pressure activated operation

VWR® mini vortex mixer provides high speed vortexing (2750 rpm) with touch activation.

Certifications: CE marked
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR