Cole-Parmer® Essentials Glass Dome Desiccators, Antylia Scientific
Supplier: Antyila Scientific
06525-41 06525-42 06525-43
76538-936EA 186.06 CAD
76538-936 76538-938 76538-940
Cole-Parmer® Essentials Glass Dome Desiccators, Antylia Scientific
Desiccators Desiccators, Plastic
Clear glass provides excellent visibility.
  • Remove cover easily with built-in knob
  • Autoclavable
Manufactured from durable borosilicate glass, these clear glass dome desiccators are designed for analytical work. The borosilicate glass lid features a molded-in knob for easy handling of the cover. To create an airtight seal, it is recommended you grease the rim and lid of the desiccator.
The included perforated porcelain desiccator plate fits onto a ridge above the lower chamber. A standard drying agent such as silica or anhydrous calcium chloride can be placed in the lower chamber.
Delivery information: Supplied with perforated porcelain desiccator plate.