Cole-Parmer® SH-200 Series Microplate Shakers, Antylia Scientific
Supplier: Antylia Scientific
51901-09 51901-17
76542-136EA 2849.95 CAD
76542-136 76542-134
Cole-Parmer® SH-200 Series Microplate Shakers, Antylia Scientific
Shakers Microplate Shakers
Provides a fast-mixing action in a tight orbit.
  • Non-slip mat and platform lip securely hold plates
  • Built-in timer and digital speed LED display provide reproducible results
  • Can be use in incubators
These microplate shakers combine high speed and a tiny 1.5 mm orbit to produce vigorous shaking perfect for microwell plates. The shakers offer a quiet drive mechanism, easy to use controller with timer, LED display, and a digital speed selection from 250 to 1250 rpm with a soft start.
Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert. Shaker is capable of continuous use, and the microprocessor maintains speed accuracy over long runs.
Choose the four-plate model for small scale applications or the eight-plate model for high throughput bench_top applications.
Delivery information: Supplied with flat platform, non-slip mat and power cord.