High Performance Muffle Furnace, FP Series, Yamato
Supplier: Yamato Scientific
FP-103 FP-303 FP-313 FP-413
76550-238EA 14454.69 CAD
76550-238 76550-240 76550-242 76550-244
High Performance Muffle Furnace, FP Series, Yamato
Ovens Furnaces (Ashing and Muffle Ovens)
Muffle furnace is designed to maintain uniform high heat temperatures and make the unit last for years. The units in this line have firmly sealed chamber doors to provide excellent heat tightness. They also come with a top-quality alumina porcelain hot plate where the heater is not exposed to the inner chamber. This ensures that no sample contamination will take place.
  • RCF-free chamber
  • High-temperature accuracy at ±1.0 °C
  • Upgraded with long-life R-thermocouple sensors
  • High accuracy controller for better operability and visibility
  • Excellent heat tightness
More Product Information