SafeMask® SofSkin® Procedure Earloop Face Masks, Medicom
Supplier: Medicom
2080 2081 2086 2087 2082 2083 2088 2089
CA76518-344CS 175.81 CAD
CA76518-344 CA76518-346 CA76518-348 CA76518-350 CA76544-822 CA76544-824 CA76544-826 CA76544-828
SafeMask® SofSkin® Procedure Earloop Face Masks, Medicom
Masks Surgical Masks
The highest level of protection in the softest mask.
  • Non-woven spunbond outer layer provides maximum protection against fluid penetration
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • High-quality media ensures optimal filtration and breathability
  • Adjustable, enclosed nosepiece forms a strong seal to provide customized protection and comfort
  • Latex-free
Made especially for sensitive skin. Lightweight, breathable inner layer softens with wear and remains cool on skin. Unique wicking technology keeps face dry and comfortable. Soft,More Product Information