AssureMask® Valiant™ Procedure Earloop Face Masks
Supplier: Medicom
205714 205715 205716 205717 205814 205815 205816 205817 205914 205915 205916 205917 205717 205814 205816 205817 205917
CA76544-778CS 92.49 CAD
CA76544-778 CA76544-780 CA76544-782 CA76544-784 CA76544-786 CA76544-788 CA76544-790 CA76544-792 CA76544-794 CA76544-796 CA76544-798 CA76544-800 77603-754 77603-756 77603-760 77603-762 77603-770
AssureMask® Valiant™ Procedure Earloop Face Masks
Masks Surgical Masks
Procedure masks are intended to be worn by personnel during medical procedures to protect both the patient and the operating personnel from the transfer of microorganisms, body fluids, and particulate material.
  • Non-woven spunbond polypropylene outer layer provides maximum protection against fluid penetration
  • High-quality media ensures optimal filtration and breathability
  • Non-woven spunbond polypropylene inner layer is made of dye-free material that will not lint or tear during use
  • Ample length ear loops are soft, strong, free of natural rubber latex, ultrasonically sealed and attached to the outside of the mask to prevent pulling and irritation
  • Adjustable, enclosed, plastic-covered, iron nosepiece provides customized protection and comfort
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • Made in North America to meet the highest quality and regulatory standards
Premium materials and over 30 years of expertise go into every Medicom AssureMask® Valiant™ mask to provide the reliable respiratory protection and optimal comfort and breathability that healthcare professionals need. Made from high-quality materials that offer healthcare providers the reliable respiratory protection they need without restricting breathing or causing irritation, even during the longest shifts.
Delivery information: Store in cool, clean, and dry conditions. Avoid excessive heat (over 40°C or 104°F).
Packaging: Masks are packaged 50/box, 10 boxes/case.