Cole-Parmer® Essentials Vacuum Desiccators with White Base, PP/PC, Antylia Scientific
Supplier: Antyila Scientific
06525-31 06525-32 06525-33 06525-34
76538-920EA 144.23 CAD
76538-920 76538-922 76538-924 76538-926
Cole-Parmer® Essentials Vacuum Desiccators with White Base, PP/PC, Antylia Scientific
Desiccators Desiccators, Plastic
Holds vacuum up to 29” Hg for 24 hours.
  • Knurled knob provides easy handling of dome
  • Autoclavable
These vacuum desiccators can hold a vacuum up to 29” (740 mm) Hg for 24 hours without any greasing. The top lid with molded-in knurled knob is constructed of rigid, transparent polycarbonate (PC), providing a clear view of your sample.
The lower half is made of white polypropylene (PP) and includes a perforated PP desiccator plate. An internal groove on the flange holds the silicon rubber O-ring that provides leakproof vacuum capability.
A PP stopcock accepts ¼” (6.4 mm) ID vacuum tubing and features a self-lubricating PTFE plug. The three-way plug allows the user to create, hold, and release the vacuum. Autoclavable at 121 °C for 15 minutes.
Delivery information: Supplied with perforated PP desiccator plate, silicon O-ring, stopcock, and three-way plug.