Purifier® Vertical Clean BenchVertical, 6', Labconco®
Supplier: Labconco
3660600 3660601 3660603 3660604 3660620 3660621 3660623 3660624
76545-610EA 22151.6 CAD
76545-610 76545-612 76545-614 76545-616 76545-618 76545-620 76545-622 76545-624
Purifier® Vertical Clean BenchVertical, 6', Labconco®
Fume Hoods and Enclosures Laminar Flow Cabinets
Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.
Used in pharmacy compounding, plant tissue culture, media plate preparation, electronics inspection, medical device assembly and more.
Certifications: Promotes ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
Accessories information: The IV Bar supports intravenous solution bottles and bags. Includes IV bar, mounting hardware, four hangers and instructions for installation.