Logic® Vue Class II Enclosures, for Protection and Containment for Large Instruments, Labconco
Supplier: Labconco
3300400 3300401 3300410 3300411 3300610 3300611
76545-592EA 57053.8 CAD
76545-592 76545-594 76545-596 76545-598 76545-600 76545-602
Logic® Vue Class II Enclosures, for Protection and Containment for Large Instruments, Labconco
Fume Hoods and Enclosures Biological Safety Cabinets
The Logic® Vue is a specialized Class II enclosure providing personnel and product protection for large automated instruments such as liquid handlers. The Logic® Vue provides laminar ISO 5 air.
  • Large capacity enclosure
  • Accommodates diverse range of instruments such as liquid handlers and cell sorters
  • Provides accessibility from front and back
  • Visibility from all sides
  • Powered by MyLogic™ OS to interface with a variety of devices for additional researcher safety
Certifications: Meets NSF/ANSI Standard 49.
Accessories information: 76545-604 CanopyMore Product Information