Masterflex® L/S® Stainless-Steel IP65 Precision Pump System, Avantor®
Supplier: Avantor Fluid Handling
77714-86 77714-87 77714-86 77714-87 77714-86 77714-87 77714-86 77714-87 77714-86 77714-87 77714-86 77714-87 77714-86 77714-87 77714-86 77714-87 77714-86 77714-87
MFLX77714-86EA 8279.2 CAD
MFLX77714-86 MFLX77714-87 MFLX77714-86-AU MFLX77714-87-AU MFLX77714-86-DK MFLX77714-87-DK MFLX77714-86-IN MFLX77714-87-IN MFLX77714-86-IL MFLX77714-87-IL MFLX77714-86-EU MFLX77714-87-EU MFLX77714-86-UK MFLX77714-87-UK MFLX77714-86-CH MFLX77714-87-CH MFLX77714-86-IT MFLX77714-87-IT MFLX77714-89
Masterflex® L/S® Stainless-Steel IP65 Precision Pump System, Avantor®
Pumps Peristaltic Pumps
Easy-to-use drives offer precise speed control and digital display of rpm—NEMA 4X housings stand up to process and washdown applications.
  • Four digit LED display shows motor rpm—confirm speed setting at a glance
  • Integrated open-head sensor stops drive motor when pump head is opened
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • Open head sensor
These accurate console drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications.
Operation of the integrated open-head sensor requires use of sensor-equipped L/S Easy-Load II pump heads 77200-30 or 77200-32.
Certifications: ETL, cETL, CE marked.
Ordering information: Includes NEMA 3-prong ground power cable. Accessories not included.