Cimarec+™ Hotplates, Thermo Scientific
Supplier: Thermo Fisher Scientific
HP88857104 HP88850100 HP88854100 HP88857100
10790-822EA 1189.27 CAD
10790-822 10790-830 10791-580 10791-586
Cimarec+™ Hotplates, Thermo Scientific
Heaters Hot Plates
The Thermo Scientific Cimarec+™ series digital hotplates are designed to provide exceptional safety, and temperature performance for routine protocols
All units offer digital displays of temperature. Temperature is adjustable in 1° increments. The HOT TOP warning system is a standard feature, and is designed to protect users from accidental burns. The HOT TOP feature provides a prominent display when heating surface is above 50 °C (122 °F), even when unit is shut off (as long as the unit remains plugged in). The Cimarec+™ series also features automatic over-temperature protection, and aMore Product Information