J&W VF-200ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP5891 CP5892 CP8855 CP8858 CP8859 CP8860 CP8865 CP8869
CAAGCP5891EA 1105.34 CAD
CAAGCP5891 CAAGCP5892 CAAGCP8855 CAAGCP8858 CAAGCP8859 CAAGCP8860 CAAGCP8865 CAAGCP8869
J&W VF-200ms Columns, Agilent Technologies
Chromatography Columns
Uniquely selective for high dipole-moment compounds such as ketones, aldehydes, nitro- or chloro-containing compounds, PAHs, unsaturated compounds, silanes, and CFCs.
  • Trifluoropropyl phase has very high temperature stability and can be used routinely up to 350 °C
  • Ultra-low bleed for trace analysis
  • 0.15 mm Int.Ø columns available for high-efficiency GC and GC/MS analyses
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W VF-200ms offers unique selectivity for compounds rich in dipole-dipole interactions, due to the electrophilic nature of the trifluoropropyl stationary phase.
The column features superior surface optimized deactivation for symmetrical peak shapes, and high inertness leads to more accurate peak identification and reliable analysis.