PLRP-S for Prep to Process, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
PL1112-1801 PL1112-1802 PL1112-6800 PL1112-6801 PL1212-1103 PL1212-3703 PL1212-6100 PL1212-6801 PL1712-6800
CAAGPL11121801EA 1706.23 CAD
CAAGPL11121801 CAAGPL11121802 CAAGPL11126800 CAAGPL11126801 CAAGPL12121103 CAAGPL12123703 CAAGPL12126100 CAAGPL12126801 CAAGPL17126800
PLRP-S for Prep to Process, Agilent Technologies
Chromatography Columns
Fully porous polystyrene/divinylbenzene media suitable for oligonucleotide and protein purification.
  • Discovery stage to multi-kg cGMP production reduces method development time
  • Chemical stability for separations, optimization, sanitation, and regeneration increases selectivity and column lifetime
  • Single batch packing of multiple columns reduces system downtime and validation costs
The PLRP-S media, rigid poly(styrene/divinylbenzene) particles, are available in a range of pore sizes for small molecule, synthetic biomolecule and macromoleculeMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Max. pressure 207 bar 207 bar 207 bar 103 bar
pH range 1-14 1-14 1-14 1-14
Temperature limit 200 °C 200 °C 200 °C 200 °C
Wetted materials