J&W PLOT PT Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19091P-K15PT CP7348PT CP7351PT CP7353PT CP7354PT CP7534PT
CAAG19091PK15PEA 3038.18 CAD
CAAG19091PK15P CAAGCP7348PT CAAGCP7351PT CAAGCP7353PT CAAGCP7354PT CAAGCP7534PT
J&W PLOT PT Columns, Agilent Technologies
Chromatography Columns
PLOT column with integral particle trap at each end to eliminate the need to connect separate traps, for leak-free GC/MS operation.
  • Enhance analytical capability in the lab using PLOT analyses with MS detection
  • No more hassles with unions or downstream filters, due to integrated particle trapping technology incorporated into the column as one continuous length of fused silica tubing
  • Reproducible flow restriction and increased stability reduce the need for method/system adjustments, and promote a steadier analytical output for higher throughput
  • Lower operating costs and less downtime associated with replacing filters and column switching valves
  • Tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W PLOT PT columns have an integral particle trap at each end, eliminating the need to connect separate traps. Operation is more convenient, with no risk of leaks. Integrated particle-trapping technology on both column ends reduces downtime and allows the use of GC/MS for detailed qualitative and quantitative analysis.