J&W DB-200 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-2032 122-2033 122-2032LTM 123-2032 123-2033 125-2032
CAAG122-2032EA 1167.91 CAD
CAAG122-2032 CAAG122-2033 CAAG1222032LTM CAAG123-2032 CAAG123-2033 CAAG125-2032
J&W DB-200 Columns, Agilent Technologies
Chromatography Columns
A mid-polarity column ideal for difficult-to-separate positional isomers.
  • Temperature limits of 300 °C isothermal and 320 °C programmed
  • Mid polarity, more polar than DB-1701 or DB-17
  • Low ECD bleed and unique selectivity
  • Close equivalent to USP phase G6
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-200 is a (35% trifluoropropyl)-methylpolysiloxane column. This mid-polarity column is ideal for difficult-to-separate positional isomers. ItMore Product Information