J&W Carbobond Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7372 CP7374
CAAGCP7372EA 2728.27 CAD
CAAGCP7372 CAAGCP7374
J&W Carbobond Columns, Agilent Technologies
Chromatography Columns
A single-column solution that delivers higher productivity for ASTM D2505.
  • Stable and robust for high repeatability of results
  • Available in bonded and PLOT versions for improved versatility and enhanced productivity
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W CarboBOND is a carbon-based bonded column for ASTM D2505, which describes the measurement of ppm CO and CO₂ in ethylene and propylene streams. In contrast to a multipacked columnMore Product Information