Biodiesel Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
123-BD01 123-BD11 123-BD34 160-BD65-5 1909BD-113
CAAG123-BD01EA 786.19 CAD
CAAG123-BD01 CAAG123-BD11 CAAG123-BD34 CAAG160-BD65-5 CAAG1909BD-113
Biodiesel Columns, Agilent Technologies
Chromatography Columns
Designed and application-optimized for the analysis of biodiesel to meet ASTM and EN testing standards.
  • Complete range of biodiesel GC columns specially designed according to EN 14105, ASTM D6584, EN 14103, EN 14110, and EN 14331 methods
  • Bonded and crosslinked
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W Biodiesel is designed and application-optimized for the analysis of biodiesel to meet ASTM and EN testing standards, including ASTM D6584, EN 14013, EN 14105, EN 14106, and EN 14110. Engineered to meet the growing demand for analysis of biofuels as supplements or replacements for petroleum-based fuels, the column provides the highest level of performance for the determination of free and total glycerin, FAMEs, and trace methanol.