J&W VF-5ht Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP9046 CP9047 CP9094
CAAGCP9046EA 1345.6 CAD
CAAGCP9046 CAAGCP9047 CAAGCP9094
J&W VF-5ht Columns, Agilent Technologies
Chromatography Columns
VF-5ht improves the analysis of high boiling compounds by exhibiting low bleed, even at high temperatures.
  • Enhanced stability improves column longevity and reduces downtime
  • Superior detector performance provides improved detection limits
  • Ultra low bleed at high temperatures for analysis of high-boiling compounds
  • Optimized sensitivity and accuracy for analysis of high molecular weight compounds
  • Identical selectivity as VF-5ms (bleed spec of 30 m × 0.25 mm column is < 5 pA at 400 °C)
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W VF-5ht improves the analysis of high boiling compounds with low-bleed performance, even at high temperatures. Based on ultra-low-bleed technology, VF-5ht provides unmatched selectivity, sensitivity, and accuracy for the analysis of high molecular weight compounds. This column has temperature limits of –60 to 400 °C.