PL-SCX PL-SCX for Biomolecules, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
PL1545-1502 PL1545-1802 PL1545-3102 PL1545-3802 PL1945-1502
CAAGPL15451502EA 1430.91 CAD
CAAGPL15451502 CAAGPL15451802 CAAGPL15453102 CAAGPL15453802 CAAGPL19451502
PL-SCX PL-SCX for Biomolecules, Agilent Technologies
Chromatography Columns
Fully porous sulfonated polymeric strong cation exchanger
  • Optimal design for effective separation of biomolecules
  • Pore sizes allow use of a range of solute sizes
  • Exceptional stability for long column lifetime
PL-SCX is a macroporous PS/DVB matrix with a very hydrophilic coating and strong cation-exchange functionality. This process is controlled to provide the optimum density of strong cation-exchange moieties for the analysis, separation and purification of a wide range of biomolecules, from small peptides to large proteins. Two pore sizes are available, 1000 and 4000 Å, to provide good mass transfer characteristics for a range of solute sizes. The 5 μm media delivers separations at higher resolution with the 30 μm media used for medium pressure liquid chromatography.