PL HFIPgel, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
PL1114-6900HFIP PL1514-1900HFIP
CAGPL11146900HEA 4075.61 CAD
CAGPL11146900H CAGPL15141900H
PL HFIPgel, Agilent Technologies
Chromatography Columns
PL HFIPgel has been developed to perform reliably in HFIP and other polar fluorinated solvents. 
  • For room-temperature analysis of nylons, polyamides, polyesters, PLLA, and PLGA
  • Reliable results and extended lifetime in HFIP and fluorinated solvents
  • Avoids excessive curvature of calibration curves, dislocations, shoulders, and poor resolution caused by HFIP
  • Optimized separation range delivers high performance with no artifacts
  • Available in 4.6 mm diameters to reduce solvent consumption
PL HFIPgel has been developed to perform reliably inMore Product Information