J&W Intuvo Polyethylene Glycol (WAX) Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-3232-INT 123-3234-INT 19091F-115-INT CP97723-INT
CAAG1223232INTEA 1501.02 CAD
CAAG1223232INT CAAG1233234INT CAAG19091F115I CAAGCP97723INT
J&W Intuvo Polyethylene Glycol (WAX) Columns, Agilent Technologies
Chromatography Columns
Advanced range of WAX columns based on the polyethylene glycol (PEG) polymer, for use with the Agilent Intuvo 9000 GC system.
  • General-purpose columns with same selectivity as DB-WAX
  • DB WAX UltraInert – Optimized inertness
  • HP-INNOWax – Highest upper temperature limits of bonded PEG phases
  • DB-WAXetr – Extended temperature range
  • DB-FFAP – Nitroterephthalic acid modified
  • Part of optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials, and gas clean filters
  • For use with the Agilent Intuvo 9000 System
Agilent offers a range of industry-proven WAX columns for use with the Intuvo 9000 GC System: DB-WAX Ultra Inert, HP-INNOWax, DB-WAXetr, and DB-FFAP. Each phase is based on the polyethylene glycol (PEG) polymer; strict control of the crosslinking and deactivation processes results in various unique phase characteristics to meet your specific analytical needs.