J&W Intuvo Low-bleed Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP8944-INT CP8982-INT CP9100-INT CP9105-INT CP9205-INT
CAAGCP8944-INTEA 1617.58 CAD
CAAGCP8944-INT CAAGCP8982-INT CAAGCP9100-INT CAAGCP9105-INT CAAGCP9205-INT
J&W Intuvo Low-bleed Columns, Agilent Technologies
Chromatography Columns
Designed for chromatography of a broad range of low-level analytes (for use with the Intuvo 9000 GC System).
  • Unique combination of polysiloxane chemistry and proprietary surface deactivation
  • General purpose columns with strict testing requirements for low bleed, inertness, selectivity, and efficiency
  • Optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials, and gas clean filters
  • For use with the Agilent Intuvo 9000 GC System
Agilent J&W Low-bleed Intuvo GC/MS columns (compatible only with the Agilent Intuvo 9000 GC System) are designed for chromatography of a broad range of low-level analytes, and generate lower bleed even at high temperatures. The unique polymer chemistry and proprietary surface deactivation of these columns contribute to their adherence to the tightest quality control specifications in the industry for bleed, inertness, selectivity, and efficiency. Agilent J&W Low-bleed Intuvo GC/MS columns utilize special surface deactivation and siloxane chemistries that enhance the chromatographic performance of siloxane polymers.