PL-SAX for Prep to Process, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
PL1151-3802 PL1251-3102 PL1751-3102
CAAGPL11513802EA 2788.99 CAD
CAAGPL11513802 CAAGPL12513102 CAAGPL17513102
PL-SAX for Prep to Process, Agilent Technologies
Chromatography Columns
Fully porous quaternary amine strong anion exchange suitable for oligonucleotide and protein purification.
  • Anion-exchange purifications over a wider pH range extend applications
  • HPLC flow rates and rapid equilibration reduce purification cycle times
  • Large pore size for improved mass transfer delivers high speed, high resolution purifications
  • 1000 Å pore size for high capacity purifications
  • 4000 Å gigaporous particles with improved mass transfer for large biomolecules and high-speed, high-resolution purifications
PL-SAX strong anion-exchange materials are extremely hydrophilic and are designed for purification of biomolecules. The materials are totally polymeric and are chemically and thermally stable over a full range of HPLC conditions. The strong anion-exchange functionalities, covalently linked to a chemically stable polymer, facilitate anion-exchange purifications over a wider pH range. This stability can be exploited for column sanitation and clean-up, and also enables the use of denaturing conditions and stabilizing/solubilizing agents, as encountered in the purification of synthetic oligonucleotides with self-complementary sequences.