J&W Intuvo Petroleum Specialty Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-4002-INT 19091S-001-INT CP7491-INT
CAAG1224002INTEA 520.09 CAD
CAAG1224002INT CAAG19091S001I CAAGCP7491-INT
J&W Intuvo Petroleum Specialty Columns, Agilent Technologies
Chromatography Columns
For challenging petrochemical and petroleum analysis applications with the Intuvo 9000 GC system.
  • 100% dimethylpolysiloxane
  • HP-PONA is configured for detailed hydrocarbon analysis of petroleum process products
  • DB-Sim-Dist is designed for rapid simulated distillation ASTM Method D7798-13
  • Optimized GC flow path when used with Agilent Ultra Inert Liners, low-bleed septa,certified vials, and gas clean filters
  • For use with the Agilent Intuvo 9000 System
Agilent offers 100% dimethylpolysiloxane Intuvo GC columns (for use with the Intuvo 9000 GC System)More Product Information