Poroshell 120 Chiral, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
683775-603 683775-604 683775-607 683975-603 683975-604 683975-607 683975-609 685775-603 685775-604 685775-607 685975-603 685975-604 685975-607 685975-609 689775-603 689775-604 689975-604
CAAG683775-603EA 4122.57 CAD
CAAG683775-603 CAAG683775-604 CAAG683775-607 CAAG683975-603 CAAG683975-604 CAAG683975-607 CAAG683975-609 CAAG685775-603 CAAG685775-604 CAAG685775-607 CAAG685975-603 CAAG685975-604 CAAG685975-607 CAAG685975-609 CAAG689775-603 CAAG689775-604 CAAG689975-604
Poroshell 120 Chiral, Agilent Technologies
Chromatography Columns
Chiral selectors on superficially porous particles for fast chiral separations in NP, RP, and SFC.
  • Outstanding performance and efficiency in fast and ultrafast chiral separations due to 2.7 µm superficially porous, 'core-shell' particles
  • Compatible with all common chiral HPLC modes including NP, RP, SFC, polar organic (PO), and polar ionic (PI)
  • Part of the InfinityLab family – designed for ease-of-use and maximum compatibility with Agilent instruments and supplies
  • Enhanced usability, traceability, and security with a preprogrammed ID tag
  • Use Poroshell 120
More Product Information