J&W HP-Chiral β Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19091G-B133 19091G-B213 19091G-B233
CAAG19091GB133EA 1225.33 CAD
CAAG19091GB133 CAAG19091GB213 CAAG19091GB233
J&W HP-Chiral β Columns, Agilent Technologies
Chromatography Columns
Permethylated ß-cyclodextrin chiral stationary phase that resolves volatile optical isomers.
  • ß-cyclodextrin in (35%-Phenyl)-methylpolysiloxane
  • Permethylated ß-cyclodextrin
  • Chiral separations without chiral-specific derivatization
  • Phenyl-based polymer provides low bleed and does not interfere with nitrogen-specific detectors
  • Available in two concentrations of ß-cyclodextrin: 10% and 20%
  • 20% ß-cyclodextrin is the best choice for initial screening
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency