J&W CP-Al₂O₃/KCl Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7515 CP7518 CP7519 CP7577
CAAGCP7515EA 2327.62 CAD
CAAGCP7515 CAAGCP7518 CAAGCP7519 CAAGCP7577
J&W CP-Al₂O₃/KCl Columns, Agilent Technologies
Chromatography Columns
Agilent J&W CP-Al₂O₃/KCl is an aluminum oxide PLOT column that offers high selectivity for separating ppm levels of C1 to C5 hydrocarbons in process streams, without the need for sub-ambient cooling.
  • High capacity thick films
  • No need for sub-ambient cooling simplifies operation
  • Choice of two selectivities covers a broad range of applications
  • Available in Fused Silica and UltiMetal
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: CP-Al₂O₃/KCl,
More Product Information