AdvanceBio Peptide Plus, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
693975-349 693975-949 695775-949 821725-954
CAAG693975-349EA 1972.55 CAD
CAAG693975-349 CAAG693975-949 CAAG695775-949 CAAG821725-954
AdvanceBio Peptide Plus, Agilent Technologies
Chromatography Columns
Quickly confirm and identify target proteins and peptides with this reversed phase c18 column.
  • Reliable results - high resolution separations delivered by efficient Poroshell particle morphology, with no deterioration in performance for higher mass loads
  • Superior peak shape - chemically modified silica with a hybrid charged C18 surface for superior performance with formic acid as the mobile phase additive
  • Alternate Selectivity – to traditional C18 columns for improved separation/resolution of critical components
  • Increased flexibility - compatibility with both HPLC and UHPLC systems via 2.7 μm, 100 Å, diameter superficially porous particles and multiple geometries
  • Quality control - columns are batch tested with peptide standards to ensure quality
AdvanceBio Peptide Plus columns are reversed phase, superficially porous particle LC columns optimized for the separation of target peptides, impurities, and post translational modifications. These columns feature a hybrid endcapped C18 stationary phase on a 100 Å pore size, 2.7 μm particle modified to have a charged surface. AdvanceBio Peptide Plus columns enable you to identify multiple critical quality attributes confidently by LC/MS. They provide the speed, sensitivity, and efficiency you need to quickly confirm and identify target proteins and peptides.