AdvanceBio Amino Acid Analysis (AAA), Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
695975-322 823750-946
CAAG695975-322EA 1565.33 CAD
CAAG695975-322 CAAG823750-946
AdvanceBio Amino Acid Analysis (AAA), Agilent Technologies
Chromatography Columns
For fast and reproducible pre-column derivatization and separation of amino acids in protein hydrolysates and cell culture media.
  • Reliable results - high-resolution separations delivered by efficient Poroshell particle morphology
  • Reduced costs - long column lifetimes from robust, high-pH resistant, chemically modified silica
  • Increased flexibility - compatibile with both HPLC and UHPLC systems via 2.7 μm diameter particles
  • Quality control - AdvanceBio AAA columns are batch tested with amino acid standards to ensure quality
  • Easy ordering - standards and reagents available as kits
  • Automated online derivatization - with Agilent analytical injection systems
AdvanceBio Amino Acid Analysis (AAA) includes proven reagents for amino acid derivatization, a ready-to-use amino acid standards kit, columns based on innovative Poroshell technology, and expert support.
AdvanceBio AAA delivers fast, sensitive, and reproducible separations of amino acids in biological samples, protein hydrolysates, and cell culture media. Used alongside Agilent InfinityLab LC Series instruments, AdvanceBio AAA provides a complete solution for amino acid analysis.