VanGuard 1400 Series Compound Microscopes, Vee Gee Scientific
Supplier: Heathrow Scientific
1423PHI 1431BRI
76546-002EA 3983.16 CAD
76546-002 76546-004 76546-014 76546-016
VanGuard 1400 Series Compound Microscopes, Vee Gee Scientific
Microscopes
Built around a full-size frame, the 1400 series eliminates the fatigue and strain associated with working long hours over a microscope.
  • Low-position focusing controls and long drop-down X-Y controls ensure simple, consistent, and comfortable use
  • Plan objectives provide exceptionally high optical flatness and visual clarity
  • Choose from binocular or trinocular viewing heads, and Brightfield or Phase Contrast configurations
Optical system: VIOS infinity corrected.
Dioptric: −6 to +6
Ordering information: 5 year optical/mechanical; 1 year electrical warranty.