Masterflex® Normally Open Flow Switch for Low Pressure-Drop Liquids, Avantor®
Supplier: VWR International
MFLX32776-00 MFLX32776-01 MFLX32776-02 MFLX32776-03
MFLX32776-00EA 330.22 CAD
MFLX32776-00 MFLX32776-01 MFLX32776-02 MFLX32776-03
Masterflex® Normally Open Flow Switch for Low Pressure-Drop Liquids, Avantor®
Flow Switches
Minimizes flow constriction.
  • Provides low pressure drop
These SPST switches feature a unique, dual-diameter, internal bore and piston configuration to minimize flow constriction and provide low pressure drop. Use a 150 um filter to protect your switch.
Delivery information: Includes 1/4" male quick connect terminals.