Masterflex® Flow Sensor Flow Controllers, Avantor®
Supplier: SEAMETRICS INC
FT440 MFLX33110-70
MFLX33110-60EA 1049.12 CAD
MFLX33110-60 MFLX33110-70
Masterflex® Flow Sensor Flow Controllers, Avantor®
Flow Controllers
Easily control, view, and store your flow rates.
  • Control your Low-Flow Impeller Sensors with these controllers
These new controllers display flow rate and total (resettable) simultaneously on two 8-digit Display LCDs. Push-button keys and menu prompts make it easy to program the controller. The 4-to-20 mA output lets you send data to a recorder or datalogger. Nonvolatile memory stores flow total in the event of power failure.
Select a 1¼4 DIN panel- or wall-mount controller depending on your application needs. Panel-mount model 33110-60 features a gasketed front panel, membrane switches, and NEMA Type 12 (IP52) enclosure. Wall-mount model 33110-70 has a clear cover and splashproof NEMA Type 4X (IP56) enclosure. Both models include a 21¼2-ft L cable with stripped ends. Order power supply for 110 VAC operation separately.