Masterflex® Standard Pump Head Tubing Loading Keys, Avantor®
Supplier: Avantor Fluid Handling
7013-90 7019-90
MFLX07013-90EA 15.01 CAD
MFLX07013-90 MFLX07019-90
Masterflex® Standard Pump Head Tubing Loading Keys, Avantor®
Puller and Inserter Tools
Ensure proper tubing alignment and installation.
  • Constructed of durable stainless steel
L/S® and I/P® Standard pump heads include tubing loading keys. A key is required to ensure proper tubing alignment and tension. Order a replacement key based on your pump head series, L/S® or I/P®.