Masterflex® Plastic Tubing Clamp Ties, Avantor®
Supplier: VWR International
CTT60R9C2 CTT60R0HSC3
MFLX95615-00EA 80.58 CAD
MFLX95615-00 MFLX95615-10
Masterflex® Plastic Tubing Clamp Ties, Avantor®
Ties
Special curved clamp tie head ensures tight seal.
  • Ideal for general-purpose, single-use applications
Perfect for flexible fuel line, water line, and vacuum line applications. Ties measure 8" L x 0.18" W; maximum diameter of 1-3/4".