Masterflex® Laboratory Software and Prepackaged Accessories, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
AG0013
MFLX78002-92CS 95.25 CAD
MFLX78002-92
Masterflex® Laboratory Software and Prepackaged Accessories, Avantor®
Software Laboratory Software
Stand-alone process operation. No need for a PC.
  • Optimize performance of your Masterflex pumps
The ProgEdit software enables the user to program individually definable dispensing and pumping sequences with a PC and to download the program to the memory of the pump drive.
ISMATEC's MCP Process dispensing pumps are ideal for safe dispensing and filling applications. Individual, application based dispensing and pumping sequences can be programmed directly into a PC (batch-programming) and subsequently downloaded (via the RS232 interface) into one of the four program memories available on the pump drive. Similar to a simple PLC-type controller, the pump and its program will provide on the spot operation as stand-alone units and freely disconnected from the PC. This makes these ISMATEC pump drives extremely suitable for fluid processing requiring accuracy, hygienic conditions, crosslinkability, reliability and expandability. Pump sold separately.
System requirements: Pentium PC or higher, Win9x or higher, Windows NT 4.0 or higher, and 3 1/2" floppy drive.