Masterflex® Reducer Fittings, Hose Barb, Straight (HDPE), Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
C4.2-5HDPE(10 PACK) C12-10HDPE(5 PACK) C2-1.5-200HDPE(10PK) C16-12HDPE(5 PACK) C8-5HDPE(100 PACK) C2-1-200HDPE-MF(10PK) C4.2-5HDPE(100 PACK) C10-6HDPE(10 PACK) C10-4HDPE(100 PACK) C10-4HDPE(10 PACK) C8-4HDPE(10 PACK) C16-10HDPE(5 PACK) C6-4HDPE(10 PACK) C2-1-200HDPE(10PK) C4-3HDPE(100 PACK) C8-6HDPE(100 PACK) C12-8HDPE-MF(10PK) C4-2HDPE(10 PACK) C3-2HDPE(10PK) C6-5HDPE(10 PACK) C16-8HDPE(5 PACK) C8-5HDPE(10 PACK) C2-1.5-200HDPE-MF(10PK C3-2HDPE-MF(10PK) C16-8HDPE-MF(10PK) C12-10HDPE(50 PACK) C4-3HDPE(10 PACK) C6-4HDPE(100 PACK) C12-6HDPE(50 PACK) C2.5-2HDPE(100 PACK) C2.5-2HDPE(10 PACK) C4-2HDPE(100 PACK) C5-4HDPE(10 PACK) C1.5-1-200HDPE(10PK) C12-8HDPE(5 PACK) C8-6HDPE(10 PACK) C6-5HDPE(100 PACK) C10-5HDPE(10 PACK) C16-12HDPE(50 PACK) C8-4HDPE(100 PACK) C5-4HDPE(100 PACK) C10-8HDPE(10 PACK) C12-6HDPE(5 PACK)
MFLX31518-33EA 37.13 CAD
MFLX31518-33 MFLX30626-33 MFLX41518-31 MFLX30626-35 MFLX30626-13 MFLX30622-11 MFLX30622-33 MFLX30626-36 MFLX30626-17 MFLX30622-13 MFLX30622-35 MFLX30626-38 MFLX30622-36 MFLX30622-15 MFLX30622-38 MFLX30622-17 MFLX30622-29 MFLX30622-07 MFLX30622-09 MFLX30616-33 MFLX30626-40 MFLX30625-96 MFLX30626-42 MFLX41518-22 MFLX30622-40 MFLX30626-24 MFLX30622-42 MFLX30622-00 MFLX30622-24 MFLX30626-05 MFLX30626-27 MFLX30622-03 MFLX41518-29 MFLX30626-29 MFLX30626-07 MFLX30622-05 MFLX30622-27 MFLX40616-01 MFLX30621-98 MFLX30621-96 MFLX30616-29 MFLX40616-05 MFLX30616-09
Masterflex® Reducer Fittings, Hose Barb, Straight (HDPE), Avantor®
Fittings and Connectors
Best for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range -65 to 150 °F (-53 to 65 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.