Masterflex® L/S® Drive Dual Voltage Power Supply, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
77310-02 77200-07
MFLX77310-02EA 2867.7 CAD
MFLX77310-02 MFLX77200-07
Masterflex® L/S® Drive Dual Voltage Power Supply, Avantor®
Power Supplies
Make the most of your Masterflex drives replacement parts and other accessories.
  • Suitable for use with 07571-00 L/S drive
This power supply is rated for 3.0 Amps at 12 VDC. The total output power is 36 watts at 12 VDC.