Masterflex® Panel-Mount Direct-Reading Variable-Area Flowmeters, 65-mm Scale, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
PMR1-010812 PMR1-010813 PMR1-010814 PMR1-010817 PMR1-010820 PMR1-010831 PMR1-010853 PMR1-010855 PMR1-010857 PMR1-010858 PMR1-010861 MFLX32010-01 MFLX32010-09 MFLX32010-11 MFLX32010-15 MFLX32010-16 MFLX32010-31 MFLX32010-35 MFLX32011-03 MFLX32011-07 MFLX32011-09 MFLX32011-13 MFLX32011-16 MFLX32011-17 MFLX32011-31 MFLX32011-33 MFLX32012-01 MFLX32012-05 MFLX32012-09 MFLX32012-15 MFLX32012-16 MFLX32012-29 MFLX32012-31 MFLX32012-35
MFLX32010-03EA 202.24 CAD
MFLX32010-03 MFLX32010-05 MFLX32010-07 MFLX32010-13 MFLX32010-17 MFLX32011-01 MFLX32012-03 MFLX32012-07 MFLX32012-11 MFLX32012-13 MFLX32012-17 MFLX32010-01 MFLX32010-09 MFLX32010-11 MFLX32010-15 MFLX32010-16 MFLX32010-31 MFLX32010-35 MFLX32011-03 MFLX32011-07 MFLX32011-09 MFLX32011-13 MFLX32011-16 MFLX32011-17 MFLX32011-31 MFLX32011-33 MFLX32012-01 MFLX32012-05 MFLX32012-09 MFLX32012-15 MFLX32012-16 MFLX32012-29 MFLX32012-31 MFLX32012-35
Masterflex® Panel-Mount Direct-Reading Variable-Area Flowmeters, 65-mm Scale, Avantor®
Flow Meters
No more eye strain - front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.
Delivery information: Includes 150-mesh inlet screen.