Masterflex® Direct-Read Variable-Area Flowmeters for Water, Polycarbonate Shield, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
F3C-F01-01-SA F3C-F02-01-TF F3C-F04-01-TF F3C-F05-01-TF F3D-F06-01-TF F3E-F10-01-TF F6C-F01-01-SA F6C-F03-01-TF F6C-F05-01-TF F6D-F06-01-TF MFLX32056-04 MFLX32056-12 MFLX32056-20 MFLX32056-22 MFLX32057-02 MFLX32057-12 MFLX32057-14 MFLX32057-16 MFLX32057-18 MFLX32057-24
MFLX32056-00EA 519.82 CAD
MFLX32056-00 MFLX32056-02 MFLX32056-06 MFLX32056-08 MFLX32056-10 MFLX32056-18 MFLX32057-00 MFLX32057-04 MFLX32057-08 MFLX32057-10 MFLX32056-04 MFLX32056-12 MFLX32056-20 MFLX32056-22 MFLX32057-02 MFLX32057-12 MFLX32057-14 MFLX32057-16 MFLX32057-18 MFLX32057-24
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Polycarbonate Shield, Avantor®
Flow Meters
Economical design includes safety features for metering the most aggressive fluids.
  • Removable polycarbonate shield provides added safety
  • Individually leak tested assures leak-proof integrity*
  • Sight tube guide rod prevents float oscillation in dynamic systems
Ideal for use in applications requiring the high purity/high temperature suitability that fluoropolymers are capable of. Common applications include those with acids, caustics, solvents, and high purity liquids.
The solid PTFE float is designed to maximize each meter's flow range so that many ofMore Product Information